Learn More
CCT5 Mouse, anti-Human, CoraLite Plus 488, Clone: 1B12E10, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | CCT5 |
|---|---|
| Clone | 1B12E10 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18030746
|
Proteintech
CL488-67400-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18085415
|
Proteintech
CL488-67400-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CCT5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCT5. This antibody recognizes human, mouse, rat antigen. The CCT5 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| CCT5 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 22948 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1B12E10 | |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat | |
| CCT epsilon, CCT5, CCTE, KIAA0098, TCP 1 epsilon | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.