Learn More
CCT5 Mouse, anti-Human, CoraLite Plus 488, Clone: 1B12E10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67400-100UL
 View more versions of this product
Product Code. 18030746
  • € 450.00 / 100µL
Description
The CCT5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCT5. This antibody recognizes human, mouse, rat antigen. The CCT5 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
CCT5
1B12E10
CoraLite Plus 488
Mouse
Primary
Human, Mouse, Rat
155 kDa
IgG1
Immunofluorescence, Flow Cytometry (Intracellular Staining)
1000 μg/mL
CCT epsilon, CCT5, CCTE, KIAA0098, TCP 1 epsilon
100 μL
22948
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.