missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD-M6PR Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB100-75286-0.025mg
This item is not returnable.
View return policy
Description
CD-M6PR Polyclonal specifically detects CD-M6PR in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence.
Specifications
| CD-M6PR | |
| Polyclonal | |
| PBS with No Preservative | |
| 000 Man6PR, 46 kDa mannose 6-phosphate receptor, 46-kDa mannose 6-phosphate receptor, cation-dependent mannose-6-phosphate receptor, CD Man-6-P receptor, CD-MPR, FLJ32994, M6PR, mannose-6-phosphate receptor (cation dependent), MPR 46, MPR46, MPRD, Mr 46, SMPR | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mannose 6 Phosphate Receptor (Cation dependent) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| P20645 | |
| M6PR | |
| The immunogen for this antibody was a sequence from 60a.a. to 114a.a. from Mannose 6 Phosphate Receptor (Cation dependent)(a 277a.a. long protein) | |
| 0.025 mg | |
| Immunology | |
| 4074 | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction