missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD-M6PR Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
€ 312.00
Specifications
| Antigen | CD-M6PR |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Chicken |
Description
CD-M6PR Polyclonal specifically detects CD-M6PR in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence.Specifications
| CD-M6PR | |
| Polyclonal | |
| Chicken | |
| Immunology | |
| PBS with No Preservative | |
| 000 Man6PR, 46 kDa mannose 6-phosphate receptor, 46-kDa mannose 6-phosphate receptor, cation-dependent mannose-6-phosphate receptor, CD Man-6-P receptor, CD-MPR, FLJ32994, M6PR, mannose-6-phosphate receptor (cation dependent), MPR 46, MPR46, MPRD, Mr 46, SMPR | |
| M6PR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P20645 | |
| 4074 | |
| The immunogen for this antibody was a sequence from 60a.a. to 114a.a. from Mannose 6 Phosphate Receptor (Cation dependent)(a 277a.a. long protein) | |
| Affinity Purified | |
| Mannose 6 Phosphate Receptor (Cation dependent) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title