Learn More
CD105 (Endoglin) Mouse, anti-Human, FITC, Clone: 43A3, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60039FI
Encompass_Preferred
 View more versions of this product
Product Code. 17168711
  • € 321.30 / 100 tests
Description
  • The 43A3 antibody reacts with CD105 (endoglin), an ∼180 kDa cell surface glycoprotein which is a disulfide-bonded homodimer of ∼90 kDa type I transmembrane subunits
  • CD105 is a component of the TGF-β receptor complex and is expressed by vascular endothelial smooth muscle cells, syncytiotrophoblasts of placenta and activated macrophages, and at relatively low levels by stromal fibroblasts
  • Its expression is also observed in some types of tumors, and levels are up-regulated on the endothelium during angiogenesis
  • CD105 has important roles in angiogenesis, cardiovascular development, and vascular remodeling, and the protein serves a regulatory role in cytoskeletal reorganization by modulating the sites of focal adhesion and cellular migration
  • This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Specifications
Specifications
CD105 (Endoglin)
Monoclonal
FITC
100 Tests
Human, Mouse
Hematopoietic Stem and Progenitor Cells, Mesenchymal Stem and Progenitor Cells
Immunoprecipitation, Western Blot, Flow Cytometry
43A3