Learn More
CD115 Rat, PE, Clone: AFS98, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB53-PE-100ug
This item is not returnable.
View return policy
Specifications
| CD115 | |
| Monoclonal | |
| 0.2mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| Csf1r | |
| Protein A/G Purified | |
| RUO | |
| 12978 | |
| IgG2a κ |
| Flow Cytometry | |
| AFS98 | |
| PE | |
| XP_017173299 | |
| Rat | |
| 100 μg | |
| Primary | |
| Store at 2-4°C. |