Learn More
CD115 Rat, PE, Clone: AFS98, enQuireBio™
Rat Monoclonal Antibody
€ 169.05 - € 301.35
Specifications
| Antigen | CD115 |
|---|---|
| Clone | AFS98 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15972178
|
EnquireBio
QAB53-PE-25ug |
25 μg |
€ 169.05
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15962178
|
EnquireBio
QAB53-PE-100ug |
100 μg |
€ 301.35
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD115 | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 12978 | |
| IgG2a κ | |
| Protein A/G Purified |
| AFS98 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| XP_017173299 | |
| Csf1r | |
| Primary | |
| Store at 2-4°C. |