missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117/c-kit Antibody (2B8) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 11 publications
Brand: Novus Biologicals NB100-77477-0.1MG
This item is not returnable.
View return policy
Description
CD117/c-kit Monoclonal specifically detects CD117/c-kit in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| CD117/c-kit | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry <= 0.6ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| CD117 (2B8) | |
| Store at 4C. Do not freeze. | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| 2B8 | |
| Unconjugated | |
| PBS (pH 7.2) with 0.09% Sodium Azide | |
| KIT | |
| Mouse bone marrow mast cells | |
| 0.1 mg | |
| B Cell Development and Differentiation Markers, Cancer, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| 3815 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction