missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117/c-kit Antibody (2B8) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 11 publications
€ 226.00 - € 517.00
Specifications
| Antigen | CD117/c-kit |
|---|---|
| Clone | 2B8 |
| Concentration | 0.5 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18222521
|
Novus Biologicals
NB100-77477SS |
0.025 mg |
€ 226.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18218594
|
Novus Biologicals
NB100-77477-0.1MG |
0.1 mg |
€ 517.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD117/c-kit Monoclonal specifically detects CD117/c-kit in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| CD117/c-kit | |
| 0.5 mg/mL | |
| Unconjugated | |
| Rat | |
| B Cell Development and Differentiation Markers, Cancer, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| PBS (pH 7.2) with 0.09% Sodium Azide | |
| 3815 | |
| Mouse bone marrow mast cells | |
| Primary | |
| Store at 4C. Do not freeze. |
| 2B8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| KIT | |
| IgG2b κ | |
| Protein A or G purified | |
| CD117 (2B8) |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title