Learn More
CD117/c-kit Antibody (9D2) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NB110-93600
 View more versions of this product
Product Code. 18710954
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
CD117/c-kit Monoclonal specifically detects CD117/c-kit in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD117/c-kit
Monoclonal
LYOPH
Western Blot 1:100-1:1000, Immunohistochemistry 1:100-1:400, Immunohistochemistry-Paraffin 1:100-1:400
P05532
KIT
Mouse CD117/c-kit (extracellular domain)
0.1 mg
Cancer, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases
3815
Mouse
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
9D2
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
Rat
Protein G purified
RUO
Primary
Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG2