Learn More
CD11c Mouse anti-Human, Rhesus, Alexa Fluor™ 647, Clone: 3.9, BD Pharmingen™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 565911
 View more versions of this product
Product Code. 15804599
  • ONLINE EXCLUSIVE PRICE
    € 113.00
    € 112.62 / Each
    Save € 0.38
    0.34% Off
Description
CD11c Monoclonal antibody specifically detects CD11c in Human,Rhesus samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD11c
Monoclonal
0.2 mg/mL
Mouse
RUO
CD11c
Refrigerate (2 to 8°C)
Flow Cytometry
3.9
Alexa Fluor 647
25 μg
Primary
Human, Rhesus Monkey
IgG1 κ