Learn More
CD11c Mouse anti-Human, Rhesus, Alexa Fluor™ 647, Clone: 3.9, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565911
This item is not returnable.
View return policy
Description
CD11c Monoclonal antibody specifically detects CD11c in Human,Rhesus samples. It is validated for Flow Cytometry.
Specifications
| CD11c | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| CD11c | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 3.9 | |
| Alexa Fluor 647 | |
| 25 μg | |
| Primary | |
| Human, Rhesus Monkey | |
| IgG1 κ |