missing translation for 'onlineSavingsMsg'
Learn More

CD11c Mouse anti-Human, Rhesus, Alexa Fluor™ 647, Clone: 3.9, BD Pharmingen™

Mouse Monoclonal Antibody

€ 112.62 - € 236.26

Specifications

Antigen CD11c
Clone 3.9
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15804599
View Documents
BD Biosciences
565911
25 μg
ONLINE EXCLUSIVE PRICE
€ 113.00
€ 112.62 / Each
Save € 0.38
0.34% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15814599
View Documents
BD Biosciences
565912
0.1 mg
ONLINE EXCLUSIVE PRICE
€ 237.00
€ 236.26 / Each
Save € 0.74
0.31% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD11c Monoclonal antibody specifically detects CD11c in Human,Rhesus samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD11c
0.2 mg/mL
Monoclonal
Mouse
Human, Rhesus Monkey
Primary
CD11c
3.9
Flow Cytometry
Alexa Fluor 647
RUO
IgG1 κ
Refrigerate (2 to 8°C)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.