Learn More
CD127 Mouse anti-Human, BB700, Clone: HIL-7R-M21, BD Horizon™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 566399
 View more versions of this product
Product Code. 15845019
  • ONLINE EXCLUSIVE PRICE
    € 222.00
    € 221.75 / Each
    Save € 0.25
    0.11% Off
Description
CD127 Monoclonal antibody specifically detects CD127 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD127
Monoclonal
Brilliant Blue 700 (BB700)
25 Tests
Primary
Refrigerate (2 to 8°C)
IgG1 κ
Flow Cytometry
HIL-7R-M21