missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD127 Mouse anti-Human, BB700, Clone: HIL-7R-M21, BD Horizon™
Mouse Monoclonal Antibody
€ 221.75 - € 481.50
Specifications
| Antigen | CD127 |
|---|---|
| Clone | HIL-7R-M21 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 700 (BB700) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15845019
|
BD Biosciences
566399 |
25 Tests |
€ 222.00 € 221.75 / Each Save € 0.25 0.11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15835019
|
BD Biosciences
566398 |
100 Tests |
€ 494.00 € 481.50 / Each Save € 12.50 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD127 Monoclonal antibody specifically detects CD127 in Human samples. It is validated for Flow Cytometry.Specifications
| CD127 | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Mouse | |
| Human | |
| Primary |
| HIL-7R-M21 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 κ | |
| Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title