Learn More
CD152 / CTLA-4 Armenian Hamster, PE-Cy7, Clone: UC10-4F10-11, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand: EnquireBio QAB58-PE7-25ug
This item is not returnable.
View return policy
Specifications
| CD152 / CTLA-4 | |
| Monoclonal | |
| 0.2mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| Ctla4 | |
| Protein A/G Purified | |
| RUO | |
| 12477 | |
| IgG |
| Flow Cytometry | |
| UC10-4F10-11 | |
| PE-Cyanine7 | |
| NP_001268905 | |
| Armenian Hamster | |
| 25 μg | |
| Primary | |
| Store at 2-4°C. |