missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD152 / CTLA-4 Armenian Hamster, PE-Cy7, Clone: UC10-4F10-11, enQuireBio™
Armenian Hamster Monoclonal Antibody
€ 131.25 - € 288.75
Specifications
| Antigen | CD152 / CTLA-4 |
|---|---|
| Clone | UC10-4F10-11 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15922238
|
EnquireBio
QAB58-PE7-25ug |
25 μg |
€ 131.25
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15912238
|
EnquireBio
QAB58-PE7-100ug |
100 μg |
€ 288.75
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD152 / CTLA-4 | |
| 0.2mg/mL | |
| Monoclonal | |
| Armenian Hamster | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 12477 | |
| IgG | |
| Protein A/G Purified |
| UC10-4F10-11 | |
| Flow Cytometry | |
| PE-Cyanine7 | |
| RUO | |
| NP_001268905 | |
| Ctla4 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title