Learn More
CD19 Rat, anti-Mouse, Unconjugated, Clone: 1D3, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60112.1
Encompass_Preferred
 View more versions of this product
Product Code. 17129111
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • The 1D3 antibody reacts with mouse CD19, an ∼95 kDa type 1 transmembrane glycoprotein expressed on the surface of B cells throughout all stages of development, from early pre-B cells to plasma cells
  • Expression is down-regulated but persists in terminally differentiated plasma cells
  • CD19 is also found on follicular dendritic cells
  • By associating with CD21 and CD81, CD19 functions as a co-receptor for the B cell receptor and is involved in B cell activation and differentiation
  • Activation of CD19 is accompanied by phosphorylation of the cytoplasmic domain, which promotes binding to kinases and the induction of intracellular signaling cascades
  • Mutations in CD19 can result in severe immunodeficiency syndromes
  • Clone 1D3 recognizes the same epitope as clone 6D5 in cross-competition assays
Specifications
Specifications
CD19
Monoclonal
Unconjugated
100 μg
Mouse
B Cells
Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
1D3
Rat
Primary
IgG2a κ