missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Rat, anti-Mouse, Unconjugated, Clone: 1D3, STEMCELL Technologies
Rat Monoclonal Antibody
POA - € 530.55
Specifications
| Antigen | CD19 |
|---|---|
| Clone | 1D3 |
| Applications | Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17129111
|
Stemcell Technologies
60112.1 |
100 μg | |||||||
17119111
|
Stemcell Technologies
60112 |
500 μg |
€ 530.55
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The 1D3 antibody reacts with mouse CD19, an ∼95 kDa type 1 transmembrane glycoprotein expressed on the surface of B cells throughout all stages of development, from early pre-B cells to plasma cells
- Expression is down-regulated but persists in terminally differentiated plasma cells
- CD19 is also found on follicular dendritic cells
- By associating with CD21 and CD81, CD19 functions as a co-receptor for the B cell receptor and is involved in B cell activation and differentiation
- Activation of CD19 is accompanied by phosphorylation of the cytoplasmic domain, which promotes binding to kinases and the induction of intracellular signaling cascades
- Mutations in CD19 can result in severe immunodeficiency syndromes
- Clone 1D3 recognizes the same epitope as clone 6D5 in cross-competition assays
Specifications
| CD19 | |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Primary |
| 1D3 | |
| Monoclonal | |
| Rat | |
| IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title