missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Antibody (TS2/18) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22537-50ug
This item is not returnable.
View return policy
Description
CD2 Monoclonal specifically detects CD2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Inhibition Activity, CyTOF-ready, Block/Neutralize.
Specifications
| CD2 | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 0.5 - 2 ug/mL, Flow Cytometry 3 - 5 ug/ 10^6 cells, Immunoprecipitation 1:10 - 1:500, Inhibition Activity 0.5 ug, CyTOF-ready, Neutralization | |
| CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunoprecipitation, Inhibition Assays, CyTOF, Neutralization | |
| TS2/18 | |
| Unconjugated | |
| P06729 | |
| CD2 | |
| Human LFA-2 (CD2) | |
| 50 ug | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 914 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction