missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Antibody (TS2/18) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 213.00 - € 507.00
Specifications
| Antigen | CD2 |
|---|---|
| Clone | TS2/18 |
| Concentration | 1.0 mg/ml |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Inhibition Assays, CyTOF, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18472922
|
Novus Biologicals
NBP2-22537-50ug |
50 ug |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18159901
|
Novus Biologicals
NBP2-22537 |
200 μg |
€ 507.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD2 Monoclonal specifically detects CD2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Inhibition Activity, CyTOF-ready, Block/Neutralize.Specifications
| CD2 | |
| 1.0 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| CD2 | |
| IgG1 | |
| Protein A purified |
| TS2/18 | |
| Western Blot, Flow Cytometry, Immunoprecipitation, Inhibition Assays, CyTOF, Neutralization | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Apoptosis, Immunology | |
| P06729 | |
| 914 | |
| Human LFA-2 (CD2) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto