missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 / MS4A1 Mouse, Clone: rIGEL/773, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 931-MSM9-P1
This item is not returnable.
View return policy
Specifications
| CD20 / MS4A1 | |
| Predominantly cell surface with some cytoplasmic | |
| rIGEL/773 | |
| Unconjugated | |
| NP_690605 | |
| Mouse | |
| 33-37kDa | |
| Protein A/G Purified | |
| RUO | |
| 931 | |
| IgG2a κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| MS4A1 | |
| Recombinant full-length human MS4A1 protein | |
| Daudi, Raji, and U266, and human lymphocytes. Lymph nodes and tonsils. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction