Learn More
Invitrogen™ CD226 (DNAM-1) Monoclonal Antibody (10E5), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
€ 3385.00 - € 14910.00
Specifications
| Antigen | CD226 (DNAM-1) |
|---|---|
| Clone | 10E5 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17141202
|
Invitrogen™
16-2261-38 |
5 mg |
€ 3385.00
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17121202
|
Invitrogen™
16-2261-025 |
25 mg |
€ 10150.00
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17131202
|
Affymetrix eBioscience
16-2261-050 |
50 mg | |||||||
Description
This 10E5 MAb reacts with mouse CD226, which is also known as DNAX accessory molecule-1 (DNAM-1). CD226 is an adhesion molecule expressed primarily on T cells, including thymocytes. The highest levels of CD226 are found on naive CD8 T cells. Moreover, expression of CD226 is regulated differentially in CD4 T cells. Under certain culture conditions, CD226 can be upregulated in Th1 and downregulated in Th2 and Th0 cells. This cell surface receptor can also be detected on natural killer cells, some B cell subsets, monocytes, and platelets. Interaction with its ligands, the poliovirus receptor (CD155) and CD112, results in adhesion and cell signaling events that promote innate and adaptive immunity, including differentiation, survival, and cytotoxicity. The 10E5 MAb has been reported to inhibit antigen-specific T cell proliferation and IFN-gamma production in vivo.
DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.Specifications
| CD226 (DNAM-1) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| adhesion glycoprotein; BC051526; Cd226; CD226 antigen; CD226 molecule; DNAM 1; DNAM1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; platelet and T-cell activation antigen 1; Pta1; T lineage-specific activation antigen 1 antigen; TLiSA1 | |
| IgG2b κ | |
| Affinity Chromatography | |
| Antibody |
| 10E5 | |
| Flow Cytometry, Functional Assay | |
| Functional Grade | |
| Rat | |
| Mouse | |
| Q8K4F0 | |
| 225825 | |
| Primary | |
| 4°C | |
| Cd226 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.