Learn More
CD226 (DNAM-1) Monoclonal Antibody (11A8.7.4), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
€ 152.00 - € 459.00
Specifications
| Antigen | CD226 (DNAM-1) |
|---|---|
| Clone | 11A8.7.4 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15913244
|
Invitrogen
25-2269-41 |
25 Tests | |||||||
|
15923244
|
Invitrogen™
25-2269-42 |
100 Tests |
€ 459.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The monoclonal antibody 11A8.7.4 reacts to DNAM-1 also known as CD226. DNAM 1 is a 65 kD receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes. The ligands for CD226 are the poliovirus receptor (PVR, CD155) and Poliovirus Related Receptor (PPR2, Nectin-2, CD112). CD226 is involved in cytolytic activity by interacting with integrins and activating protein tyrosine kinase Fyn. The 11A8.7.4 antibody has been shown to increase T and NK cell-mediated cytotoxicity. Applications Reported: This 11A8.7.4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 11A8.7.4 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No.
DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.Specifications
| CD226 (DNAM-1) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q15762 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| 11A8.7.4 | |
| Flow Cytometry | |
| PE-Cyanine7 | |
| Mouse | |
| Human | |
| 10666 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.