missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD229 Recombinant Rabbit Monoclonal Antibody (001), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552397
This item is not returnable.
View return policy
Description
CD229 (Ly9) is a cell surface receptor of the CD150 family, which includes also e.g. CD48 and CD224. Receptors of this family regulate cytokine production and cytotoxicity of lymphocytes and NK cells. High levels of CD229 are found on T and B cells, where its expression increases during their maturation. It is absent on granulocytes, bone marrow-derived dendritic cells, platelets and erythrocytes. CD229 has been also reported on mouse monocytes and NK cells. CD229 interacts homophilically through its N-terminal domain and localizes to the contact site between T cells and antigen presenting B cells during antigen-dependent immune synapse formation.
Specifications
| CD229 | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with no preservative | |
| Q01965 | |
| Ly9 | |
| Recombinant Mouse CD229 Protein (Lys 48-Arg453). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA | |
| 001 | |
| Unconjugated | |
| Ly9 | |
| AI893573; CD229; CDABP0070; Cell surface molecule Ly-9; hly9; Lgp100; LY9; Ly-9; Lymphocyte antigen 9; mLY9; RP11-312J18.1; Signaling lymphocytic activation molecule 3; SLAM family member 3; SLAMF3; T100; T-lymphocyte surface antigen Ly-9 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 17085 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction