missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD229 Recombinant Rabbit Monoclonal Antibody (001), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 86.25 - € 412.00
Specifications
| Antigen | CD229 |
|---|---|
| Clone | 001 |
| Concentration | 100 μg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
30019237
![]() |
Invitrogen™
MA552397 |
100 μL |
€ 412.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019965
|
Invitrogen™
MA552398 |
20 μL |
€ 86.25
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
CD229 Recombinant Monoclonal antibody specifically detects CD229 in Mouse samples. It is validated for ELISATekniske data
| CD229 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| AI893573; CD229; CDABP0070; Cell surface molecule Ly-9; hly9; Lgp100; LY9; Ly-9; Lymphocyte antigen 9; mLY9; RP11-312J18.1; Signaling lymphocytic activation molecule 3; SLAM family member 3; SLAMF3; T100; T-lymphocyte surface antigen Ly-9 | |
| Recombinant Mouse CD229 Protein (Lys 48-Arg453). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Ly9 |
| 001 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q01965 | |
| 17085 | |
| IgG | |
| Protein A | |
| Antibody |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel