missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD23/Fc epsilon RII Antibody (SP23), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NB120-16702-100uL
This item is not returnable.
View return policy
Description
CD23/Fc epsilon RII Monoclonal antibody specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CD23/Fc epsilon RII | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant | |
| Rabbit | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Supernatant |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| SP23 | |
| Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50 | |
| BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor | |
| Recombinant fragment, corresponding to amino acids 48-248 of Human CD23. | |
| 100 μL | |
| Adaptive Immunity, Cancer, Immunology | |
| 2208 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction