missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD23/Fc epsilon RII Antibody (SP23), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 397.00 - € 1275.00
Specifications
| Antigen | CD23/Fc epsilon RII |
|---|---|
| Clone | SP23 |
| Dilution | Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18510583
|
Novus Biologicals
NB120-16702-100uL |
100 μL |
€ 397.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18529212
|
Novus Biologicals
NB120-16702-500uL |
500 μL |
€ 1275.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD23/Fc epsilon RII Monoclonal antibody specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CD23/Fc epsilon RII | |
| Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Human | |
| BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor | |
| Recombinant fragment, corresponding to amino acids 48-248 of Human CD23. | |
| Primary | |
| Store at 4°C. |
| SP23 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cancer, Immunology | |
| Tissue culture supernatant | |
| 2208 | |
| IgG | |
| Tissue culture supernatant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title