missing translation for 'onlineSavingsMsg'
Learn More

CD23/Fc epsilon RII Antibody (SP23), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NB120-16702-500uL

 View more versions of this product

Product Code. 18529212

  • € 1275.00 / 500µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD23/Fc epsilon RII Monoclonal antibody specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD23/Fc epsilon RII
Monoclonal
Unconjugated
Tissue culture supernatant
Rabbit
Tissue culture supernatant
RUO
Primary
Human
Supernatant
Immunohistochemistry, Immunohistochemistry (Paraffin)
SP23
Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50
BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Recombinant fragment, corresponding to amino acids 48-248 of Human CD23.
500 μL
Adaptive Immunity, Cancer, Immunology
2208
Store at 4°C.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.