missing translation for 'onlineSavingsMsg'
Learn More

CD24 Rat, V450, Clone: M1/69, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB33-V450-25ug

 View more versions of this product

Product Code. 15931858

  • € 196.35 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

CD24
Monoclonal
0.2mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Cd24a
Protein A/G Purified
RUO
12484
IgG2b κ
Flow Cytometry
M1/69
Violet Fluor 450 (V450)
NP_033976
Rat
25 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.