Learn More
CD27 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: M-T271, BD Horizon™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 566449
 View more versions of this product
Product Code. 15865059
  • ONLINE EXCLUSIVE PRICE
    € 423.00
    € 412.43 / Each
    Save € 10.57
    2% Off
Description
CD27 Monoclonal antibody specifically detects CD27 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD27
Monoclonal
Brilliant Blue 700 (BB700)
100 Tests
Primary
Refrigerate (2 to 8°C)
IgG1 κ
Flow Cytometry
M-T271
Mouse
RUO
Human, Baboon, Cynomolgus, Rhesus Macaque