missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: M-T271, BD Horizon™
Mouse Monoclonal Antibody
€ 214.97 - € 412.43
Specifications
| Antigen | CD27 |
|---|---|
| Clone | M-T271 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 700 (BB700) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15875059
|
BD Biosciences
566450 |
25 Tests |
€ 217.00 € 214.97 / Each Save € 2.03 0.94% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15865059
|
BD Biosciences
566449 |
100 Tests |
€ 423.00 € 412.43 / Each Save € 10.57 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD27 Monoclonal antibody specifically detects CD27 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.Specifications
| CD27 | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Mouse | |
| Human, Baboon, Cynomolgus, Rhesus Macaque | |
| Primary |
| M-T271 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 κ | |
| Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title