missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD279 (PD-1) Monoclonal Antibody (J43), Brilliant Ultra Violet™ 563, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody
€ 203.00 - € 380.00
Specifications
| Antigen | CD279 (PD-1) |
|---|---|
| Clone | J43 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18476688
![]() |
Invitrogen™
365-9985-82 |
100 μg |
€ 380.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18476708
|
Invitrogen™
365-9985-80 |
25 μg |
€ 203.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Flow CytometrySpecifications
| CD279 (PD-1) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Armenian Hamster | |
| Mouse | |
| Q02242 | |
| 18566 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| J43 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 563 | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2 | |
| IgG | |
| Affinity chromatography | |
| Pdcd1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title