missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2F-10/SLAMF9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12603-50ul
This item is not returnable.
View return policy
Description
CD2F-10/SLAMF9 Polyclonal antibody specifically detects CD2F-10/SLAMF9 in Human samples. It is validated for Western Blot
Specifications
| CD2F-10/SLAMF9 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CD2 family member 10, CD2F10, CD2F-10MGC88194, CD84 homolog 1, CD84H1, CD84-H1cluster of differentiation 2 antigen family member 10, cluster of differentiation 84 homolog 1, SF2001, signaling lymphocytic activation molecule family member 2001, signaling lymphocytic activation molecule family member 9, SLAM family member 9 | |
| Produced in rabbits immunized with E. coli-derived Human CD2F-10/SLAMF9 fragment | |
| 50 μg | |
| Immunology | |
| 89886 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction