missing translation for 'onlineSavingsMsg'
Learn More

CD2F-10/SLAMF9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 196.35 - € 304.50

Specifications

Antigen CD2F-10/SLAMF9
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18673497
View Documents
Novus Biologicals
NBP3-12603-100ul
100 μg
ONLINE EXCLUSIVE PRICE
€ 322.00
€ 304.50 / 100µL
Save € 17.50
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18633168
View Documents
Novus Biologicals
NBP3-12603-50ul
50 μg
ONLINE EXCLUSIVE PRICE
€ 208.00
€ 196.35 / 50µL
Save € 11.65
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD2F-10/SLAMF9 Polyclonal antibody specifically detects CD2F-10/SLAMF9 in Human samples. It is validated for Western Blot
Specifications

Specifications

CD2F-10/SLAMF9
Western Blot
Unconjugated
Rabbit
Immunology
PBS, pH7.0
89886
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human
CD2 family member 10, CD2F10, CD2F-10MGC88194, CD84 homolog 1, CD84H1, CD84-H1cluster of differentiation 2 antigen family member 10, cluster of differentiation 84 homolog 1, SF2001, signaling lymphocytic activation molecule family member 2001, signaling lymphocytic activation molecule family member 9, SLAM family member 9
Produced in rabbits immunized with E. coli-derived Human CD2F-10/SLAMF9 fragment
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.