missing translation for 'onlineSavingsMsg'
Learn More

CD2F-10/SLAMF9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-12603-100ul

 View more versions of this product

Product Code. 18673497

  • € 322.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD2F-10/SLAMF9 Polyclonal antibody specifically detects CD2F-10/SLAMF9 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD2F-10/SLAMF9
Polyclonal
Western Blot 1:500-1:2000
CD2 family member 10, CD2F10, CD2F-10MGC88194, CD84 homolog 1, CD84H1, CD84-H1cluster of differentiation 2 antigen family member 10, cluster of differentiation 84 homolog 1, SF2001, signaling lymphocytic activation molecule family member 2001, signaling lymphocytic activation molecule family member 9, SLAM family member 9
Produced in rabbits immunized with E. coli-derived Human CD2F-10/SLAMF9 fragment
100 μg
Immunology
89886
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS, pH7.0
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.