Learn More
CD31 Mouse anti-Human, PE, Clone: WM59, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555446
Description
The WM59 monoclonal antibody specifically binds to a 130 kDa glycoprotein, gpIIa', also known as platelet endothelial cell adhesion molecule-1, (PECAM-1). CD31 has wide tissue distribution and is expressed on platelets, monocytes, granulocytes, and in high amounts on endothelial cells. This molecule has been implicated in a number of cellular phenomena, including vascular wound healing and angiogenesis, and transendothelial migration of leukocytes in inflammatory responses.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD31 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| WM59 | |
| EndoCAM; GPIIA'; PECA1; PECAM1; Platelet endothelial cell adhesion molecule | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.