Learn More
CD31 Mouse anti-Human, PE, Clone: WM59, BD
Mouse Monoclonal Antibody
€ 56.01 - € 229.13
Specifications
| Antigen | CD31 |
|---|---|
| Clone | WM59 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15840659
|
BD Biosciences
560983 |
25 Tests |
€ 56.75 € 56.01 / Each Save € 0.74 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15808178
|
BD Biosciences
555446 |
100 Tests |
€ 242.00 € 229.13 / Each Save € 12.87 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The WM59 monoclonal antibody specifically binds to a 130 kDa glycoprotein, gpIIa', also known as platelet endothelial cell adhesion molecule-1, (PECAM-1). CD31 has wide tissue distribution and is expressed on platelets, monocytes, granulocytes, and in high amounts on endothelial cells. This molecule has been implicated in a number of cellular phenomena, including vascular wound healing and angiogenesis, and transendothelial migration of leukocytes in inflammatory responses.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD31 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| WM59 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| EndoCAM; GPIIA'; PECA1; PECAM1; Platelet endothelial cell adhesion molecule |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.