Learn More
CD35 Antibody (HB-8592) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-09019-0.2mg
 View more versions of this product
Product Code. 18610169
  • ONLINE EXCLUSIVE PRICE
    € 680.00
    € 642.60 / 0.20mg
    Save € 37.40
    5% Off
Description
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD35
Monoclonal
1 mg/mL
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Flow Cytometry
HB-8592
Unconjugated
complement component (3b/4b) receptor 1 (Knops blood group), KN
Animals were immunized with purified CR1 from human erythrocytes. Spleen cells were fused with P3X63Ag8.653 myeloma cells.
0.2 mg
Immunology
1378
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ