missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (HB-8592) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 250.00 - € 642.60
Specifications
| Antigen | CD35 |
|---|---|
| Clone | HB-8592 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619368
|
Novus Biologicals
NBP3-09019-0.025mg |
0.025 mg |
€ 250.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610169
|
Novus Biologicals
NBP3-09019-0.2mg |
0.2 mg |
€ 680.00 € 642.60 / 0.20mg Save € 37.40 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Flow CytometrySpecifications
| CD35 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Animals were immunized with purified CR1 from human erythrocytes. Spleen cells were fused with P3X63Ag8.653 myeloma cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| HB-8592 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| PBS | |
| 1378 | |
| IgG1 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title