Learn More
CD38 Mouse anti-Human, BB700, Clone: HIT2, BD Horizon™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 566446
 View more versions of this product
Product Code. 15835059
  • ONLINE EXCLUSIVE PRICE
    € 231.00
    € 223.94 / Each
    Save € 7.06
    3% Off
Description
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD38
Monoclonal
Brilliant Blue 700 (BB700)
25 Tests
Primary
Human
IgG1 κ
Flow Cytometry
HIT2
Mouse
RUO
CD38
Refrigerate (2 to 8°C)