missing translation for 'onlineSavingsMsg'
Learn More

CD38 Mouse anti-Human, BB700, Clone: HIT2, BD Horizon™

Mouse Monoclonal Antibody

€ 223.94 - € 409.01

Specifications

Antigen CD38
Clone HIT2
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Blue 700 (BB700)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15835059
View Documents
BD Biosciences
566446
25 Tests
ONLINE EXCLUSIVE PRICE
€ 231.00
€ 223.94 / Each
Save € 7.06
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15825059
View Documents
BD Biosciences
566445
100 Tests
ONLINE EXCLUSIVE PRICE
€ 434.00
€ 409.01 / Each
Save € 24.99
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD38
Flow Cytometry
Brilliant Blue 700 (BB700)
RUO
IgG1 κ
Refrigerate (2 to 8°C)
HIT2
Monoclonal
Mouse
Human
Primary
CD38
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.