missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (GK1.5) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-26585
This item is not returnable.
View return policy
Description
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface), Cell depletion, Inhibition Activity, CyTOF-ready.
Specifications
| CD4 | |
| Monoclonal | |
| 0.5mg/mL | |
| Flow Cytometry 2.5 ug 1 million cells, Immunohistochemistry 1-25 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface) 2.5 ug 1 million cells, Cell depletion, Inhibition Activity, CyTOF-ready | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Mouse. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG κ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| GK1.5 | |
| Unconjugated | |
| NP_000607 | |
| CD4 | |
| The mouse cytotoxic T lymphocyte line V4 was used as the immunogen for the CD4 antibody. | |
| 0.1 mg | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| 920 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction