missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (GK1.5) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
€ 210.00 - € 549.00
Specifications
| Antigen | CD4 |
|---|---|
| Clone | GK1.5 |
| Concentration | 0.5mg/mL |
| Dilution | Flow Cytometry 2.5 ug 1 million cells, Immunohistochemistry 1-25 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface) 2.5 ug 1 million cells, Cell depletion, Inhibition Activity, CyTOF-ready |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18451242
|
Novus Biologicals
NBP2-26585-25ug |
25 ug |
€ 210.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18422922
|
Novus Biologicals
NBP2-26585-1mg |
1 mg |
€ 413.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18123603
|
Novus Biologicals
NBP2-26585 |
0.1 mg |
€ 549.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface), Cell depletion, Inhibition Activity, CyTOF-ready.Specifications
| CD4 | |
| 0.5mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| NP_000607 | |
| 920 | |
| The mouse cytotoxic T lymphocyte line V4 was used as the immunogen for the CD4 antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| GK1.5 | |
| Flow Cytometry 2.5 ug 1 million cells, Immunohistochemistry 1-25 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface) 2.5 ug 1 million cells, Cell depletion, Inhibition Activity, CyTOF-ready | |
| Unconjugated | |
| Rat | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| CD4 | |
| IgG κ | |
| Protein G purified | |
| This product reacts with Mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title