missing translation for 'onlineSavingsMsg'
Learn More

CD45.2 Mouse, FITC, Clone: 104, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB43-F-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942028

  • € 157.00 / 100µg

Estimated Shipment: 13-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

CD45.2
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Ptprc
Protein A/G Purified
RUO
19264
IgG2a κ
Flow Cytometry
104
FITC
NP_001255215
Mouse
100 μg
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers