missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD45RB Monoclonal Antibody (16A), FITC
Rat Monoclonal Antibody
€ 228.00 - € 485.00
Specifications
| Antigen | CD45RB |
|---|---|
| Clone | 16A |
| Concentration | 0.1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15324161
|
Invitrogen™
MA517884 |
100 μg |
€ 228.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15334161
|
Invitrogen™
MA517885 |
300 μg |
€ 485.00
300µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45RB Monoclonal Antibody for Flow
CD45RB is a receptor-type protein tyrosine phosphatase, CD45 glycoprotein. CD45 is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases, promotes cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45 isoforms differ in their extracellular domains, but share identical transmembrane and cytoplasmic domains. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45RB is expressed in microglia and inflammatory cells. In addition to involvement in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is involved in DNA fragmentation during apoptosis. CD45 is expressed on cells of the human hematopoietic lineage with the exception of mature red cells.Specifications
| CD45RB | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| FITC | |
| Rat | |
| RUO | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| P06800 | |
| 19264 | |
| TH2 cell clones, final boost with TH2 clone D10 Fusion Partner: Ag8.653. | |
| Antibody |
| 16A | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Mouse | |
| Ptprc | |
| B220; CD45; CD45 antigen; CD45R; GP180; LCA; L-CA; Leukocyte common antigen; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen | |
| Ptprc | |
| Primary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title