missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RC Antibody (OX22) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB120-6406-0.025MG
This item is not returnable.
View return policy
Description
CD45RC Monoclonal specifically detects CD45RC in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CD45RC | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000 | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB120-6406 recognizes rat CD45RC, the high molecular weight form of the leucocyte common antigen. This antigen is found on B cells, approximately 50% of bone marrow cells, all CD8+ve T cells, but splits CD4+ve T cells into two populations,CD4+ CD45RC (Th 1-like) and CD4+ CD45RC low (Th-2 like). This product is routinely tested in flow cytometry on rat splenocytes. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
| OX22 | |
| Unconjugated | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| PTPRC | |
| Rat lymphocytes stimulated in culture with phytohaemagglutinin. | |
| 0.025 mg | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 5788 | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction