missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RC Antibody (OX22) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
€ 204.00 - € 380.00
Specifications
| Antigen | CD45RC |
|---|---|
| Clone | OX22 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18290755
|
Novus Biologicals
NB120-6406-0.025MG |
0.025 mg |
€ 204.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18207460
|
Novus Biologicals
NB120-6406 |
0.1 mg |
€ 380.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45RC Monoclonal specifically detects CD45RC in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CD45RC | |
| 1.0 mg/mL | |
| Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| PTPRC | |
| IgG1 | |
| Protein G purified | |
| NB120-6406 recognizes rat CD45RC, the high molecular weight form of the leucocyte common antigen. This antigen is found on B cells, approximately 50% of bone marrow cells, all CD8+ve T cells, but splits CD4+ve T cells into two populations,CD4+ CD45RC (Th 1-like) and CD4+ CD45RC low (Th-2 like). This product is routinely tested in flow cytometry on rat splenocytes. |
| OX22 | |
| Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Rat | |
| 5788 | |
| Rat lymphocytes stimulated in culture with phytohaemagglutinin. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title