missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD13 Monoclonal Antibody (OTI1A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808802S
This item is not returnable.
View return policy
Description
KCTD13 Monoclonal antibody specifically detects KCTD13 in Human samples. It is validated for Western BlotSpecifications
| KCTD13 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WZ19 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 253980 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1A4 | |
| Unconjugated | |
| KCTD13 | |
| 1500003N18Rik; AV259508; BACURD1; BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 1; BTB/POZ domain-containing protein KCTD13; CTD-2574D22.4; FKSG86; hBACURD1; Kctd13; PDIP1; PDIP1alpha; Poldip1; Polymerase delta-interacting protein 1; potassium channel tetramerisation domain containing 13; potassium channel tetramerization domain containing 13; PP6832; TNFAIP1-like protein; tumor necrosis factor alpha-inducible protein PDIP1; Unknown (protein for MGC:134443) | |
| Full length human recombinant protein of human KCTD13 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction