missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD68 Recombinant Mouse Monoclonal Antibody (KP-1)
Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544196
This item is not returnable.
View return policy
Description
CD68 Recombinant Monoclonal Antibody for IHC (P), Flow
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predominantly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human malignancies, including several Acute Myeloid Leukemia studies.
Specifications
| CD68 | |
| Recombinant Monoclonal | |
| 50 μg/mL | |
| PBS with 0.1% BSA and 0.09% sodium azide; pH 6.8-7.4 | |
| P34810 | |
| Cd68 | |
| Lysosomal Fraction Of Human Lung Macrophages. | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin) | |
| KP-1 | |
| Unconjugated | |
| Cd68 | |
| CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen | |
| Mouse | |
| Purified | |
| RUO | |
| 968 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction