missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79b (B-Cell Marker) Mouse, Clone: IGB/1844, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 974-MSM4-P1
This item is not returnable.
View return policy
Specifications
| CD79b (B-Cell Marker) | |
| Cell surface | |
| IGB/1844 | |
| Unconjugated | |
| Mouse | |
| 39kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 974 | |
| IgG1 κ |
| Immunohistochemistry, ELISA | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human CD79b protein fragment (apprx. aa29-159) | |
| Daudi or Ramos cells. Germinal center B-cells in a lymph node or tonsil. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction